Läs mer
2-Bromo-6-methoxy-1,3-benzothiazole, TRC
Produktkod. 30348699
Change view
Kvantitet:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30348699 500 mg
30330645 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30348699 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB696618500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Bromo-6-methoxy-1,3-benzothiazole
CAS 2941-58-4
Molekylformel C8 H6 Br N O S
Kvantitet 500 mg
InChI formel InChI=1S/C8H6BrNOS/c1-11-5-2-3-6-7(4-5)12-8(9)10-6/h2-4H,1H3
LEDER COc1ccc2nc(Br)sc2c1
IUPAC-namn 2-bromo-6-methoxy-1,3-benzothiazole
Molekylvikt (g/mol) 244.108
Formel vikt 242.935