missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-methyl-3-nitropyridine, 98%, Thermo Scientific Chemicals

Catalog Number 15472419
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
374633-31-5
C6H5BrN2O2
217.022
MFCD03095212
IHHLJCDGOSZFTB-UHFFFAOYSA-N
2-bromo-3-nitro-6-methylpyridine, 2-bromo-3-nitro-6-picoline, 2-bromo-6-methyl-3-nitro-pyridine, 2-bromo-3-nitro-6-methyl pyridine, pubchem5954, acmc-209it3, ksc496c9p, 6-bromo-5-nitro-2-picoline, pyridine, 2-bromo-6-methyl-3-nitro, 2-bromo-6-methyl-3-nitropyridinemethyl 6-bromomethyl nicotinate
24728013
2-bromo-6-methyl-3-nitropyridine
CC1=NC(=C(C=C1)[N+](=O)[O-])Br
This item is not returnable. View return policy
374633-31-5
C6H5BrN2O2
217.022
MFCD03095212
IHHLJCDGOSZFTB-UHFFFAOYSA-N
2-bromo-3-nitro-6-methylpyridine, 2-bromo-3-nitro-6-picoline, 2-bromo-6-methyl-3-nitro-pyridine, 2-bromo-3-nitro-6-methyl pyridine, pubchem5954, acmc-209it3, ksc496c9p, 6-bromo-5-nitro-2-picoline, pyridine, 2-bromo-6-methyl-3-nitro, 2-bromo-6-methyl-3-nitropyridinemethyl 6-bromomethyl nicotinate
24728013
2-bromo-6-methyl-3-nitropyridine
CC1=NC(=C(C=C1)[N+](=O)[O-])Br

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Color White
Quantity 1 g
Beilstein 138200
Formula Weight 217.02
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-6-methyl-3-nitropyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'