Learn More
2-Bromo-6-nitrophenol, 97%, Thermo Scientific™
Catalog Number p-7077284
Click to view available options
Quantity:
5 g
25 g
13073-25-1
C6H4BrNO3
218.01
MFCD02683216
VEJSIOPQKQXJAT-UHFFFAOYSA-N
2-bromo-6-nitrophenol, phenol, 2-bromo-6-nitro, 2-bromo-6-nitro-phenol, 6-bromo-2-nitrophenol, 6-nitro-2-bromophenol, 3-bromo-2-hydroxynitrobenzene, pubchem3114, 2-brom-6-nitrophenol, 2-nitro-6-bromophenol, acmc-209bjb
13545453
2-bromo-6-nitrophenol
OC1=C(Br)C=CC=C1[N+]([O-])=O
13073-25-1
C6H4BrNO3
218.01
MFCD02683216
VEJSIOPQKQXJAT-UHFFFAOYSA-N
2-bromo-6-nitrophenol, phenol, 2-bromo-6-nitro, 2-bromo-6-nitro-phenol, 6-bromo-2-nitrophenol, 6-nitro-2-bromophenol, 3-bromo-2-hydroxynitrobenzene, pubchem3114, 2-brom-6-nitrophenol, 2-nitro-6-bromophenol, acmc-209bjb
13545453
2-bromo-6-nitrophenol
OC1=C(Br)C=CC=C1[N+]([O-])=O
Melting Point 67.0°C to 69.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 218.01
Percent Purity 97%
Chemical Name or Material 2-Bromo-6-nitrophenol
compliance-icons
Product Identifier
  • 2-BROMO-6-NITROPHENOL, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.