missing translation for 'onlineSavingsMsg'
Läs mer

2-Bromo-9,9-dioctyl Fluorene, TRC

Produktkod. 30334983
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30334983 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30334983 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB678860100MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 2-Bromo-9,9-dioctyl Fluorene
CAS 302554-80-9
Molekylformel C29H41Br
Kvantitet 100 mg
InChI formel InChI=1S/C29H41Br/c1-3-5-7-9-11-15-21-29(22-16-12-10-8-6-4-2)27-18-14-13-17-25(27)26-20-19-24(30)23-28(26)29/h13-14,17-20,23H,3-12,15-16,21-22H2,1-2H3
LEDER CCCCCCCCC1(CCCCCCCC)c2ccccc2c3ccc(Br)cc13
IUPAC-namn 2-bromo-9,9-dioctylfluorene
Molekylvikt (g/mol) 469.54
Formel vikt 468.2392
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.