missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromoacetyl)-6-methoxynaphthalene, 96%, ACROS Organics™

Brand:  Acros Organics 450060010

Additional Details : CAS Number : 10262-65-4 Weight : 0.00100kg Transport : UN number : 3261 Chem class : 8 Pack group : II


Quantity 1g

 View more versions of this product

Product Code. 15219305

  • 895.65 SEK / 1g


Estimated Shipment
09-11-2020
Add to basket


Specifications

Chemical Identifiers

10262-65-4
279.13
2-bromoacetyl-6-methoxynaphthalene, 2-bromo-1-6-methoxy-2-naphthyl ethanone, 2-bromo-6'-methoxy-2'-acetonaphthone, 2-bromo-1-6-methoxynaphthalen-2-yl ethan-1-one, 2-bromo-1-6-methoxynaphthalen-2-yl ethanone, ethanone, 2-bromo-1-6-methoxy-2-naphthalenyl, enamine_005243, 2-bromo-1-6-methoxy-naphthalen-2-yl-ethanone
2-bromo-1-(6-methoxynaphthalen-2-yl)ethanone
C13H11BrO2
HHHKEQGAGUAOQI-UHFFFAOYSA-N
193452
COC1=CC2=C(C=C1)C=C(C=C2)C(=O)CBr

Specifications

2-(Bromoacetyl)-6-methoxynaphthalene
112.0°C
10262-65-4
C13H11BrO2
Solubility in water: insoluble
COC1=CC2=C(C=C1)C=C(C=C2)C(=O)CBr
279.13
279.13
Glass Bottle
1g
95% min. (HPLC)
2-bromoacetyl-6-methoxynaphthalene, 2-bromo-1-6-methoxy-2-naphthyl ethanone, 2-bromo-6'-methoxy-2'-acetonaphthone, 2-bromo-1-6-methoxynaphthalen-2-yl ethan-1-one, 2-bromo-1-6-methoxynaphthalen-2-yl ethanone, ethanone, 2-bromo-1-6-methoxy-2-naphthalenyl, enamine_005243, 2-bromo-1-6-methoxy-naphthalen-2-yl-ethanone
HHHKEQGAGUAOQI-UHFFFAOYSA-N
2-bromo-1-(6-methoxynaphthalen-2-yl)ethanone
193452
96%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-(Bromoacetyl)-6-methoxynaphthalene

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.

Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P390-Absorb spillage to prevent material damage.

Documents