missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoanthracene, 98%, Acros Organics™

Brand:  Acros Organics 454880050

Additional Details : CAS Number : 7321-27-9 Weight : 0.00500kg


Quantity 5g

 View more versions of this product

Product Code. 15274408

  • 6224.93 SEK / 5g


Estimated Shipment date
from Supplier 05-03-2019
Add to basket


Specifications

Chemical Identifiers

7321-27-9
257.13
2-bromo-anthracene, anthracene, 2-bromo, pubchem19987, acmc-209opw, ksc377a5l
2-bromoanthracene
C14H9Br
PYXBCVWIECUMDW-UHFFFAOYSA-N
12346099
C1=CC=C2C=C3C=C(C=CC3=CC2=C1)Br

Specifications

2-Bromoanthracene
220.0°C to 221.5°C
7321-27-9
2-bromo-anthracene, anthracene, 2-bromo, pubchem19987, acmc-209opw, ksc377a5l
C1=CC=C2C=C3C=C(C=CC3=CC2=C1)Br
257.13
257.13
Authentic
5g
C14H9Br
PYXBCVWIECUMDW-UHFFFAOYSA-N
2-bromoanthracene
12346099
≥97.5% (GC)
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Bromoanthracene, 98%

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.