missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromobenzyl alcohol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 78.0°C to 82.0°C |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Linear Formula | BrC6H4CH2OH |
Quantity | 10 g |
Beilstein | 06, 445 |
Solubility Information | Solubility in water: slightly soluble. Other solubilities: soluble in most organic solvents |
Formula Weight | 187.04 |
Show More |
Safety and Handling
Product Identifier
- 2-Bromobenzyl alcohol
- Warning
- Acute toxicity Category 4
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'