missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromobenzyl chloride, 97%, Thermo Scientific Chemicals
280.08 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
2-Bromobenzyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromobenzyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
2-Bromobenzyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
Specifications
Boiling Point | 110°C to 111°C (15 mmHg) |
Refractive Index | 1.586 |
Quantity | 1 g |
UN Number | UN3265 |
Beilstein | 471701 |
Solubility Information | Not miscible or difficult to mix with water. |
Formula Weight | 205.49 |
Percent Purity | 97% |
Chemical Name or Material | 2-Bromobenzyl chloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'