Learn More
2-Bromomethyl-1,3-dioxolane, 97%, Thermo Scientific Chemicals
Catalog Number p-7024515
Click to view available options
Quantity:
25 g
100 g
500 g
4360-63-8
C4H7BrO2
167.002
MFCD00003214
CKIIJIDEWWXQEA-UHFFFAOYSA-N
2-bromomethyl-1,3-dioxolane, bromoacetaldehyde ethylene acetal, 1,3-dioxolane, 2-bromomethyl, 1,3-dioxolan-2-yl methyl bromide, bromomethyl dioxolane, acmc-1afrf, 2-bromomethyl-1,3-dioxolan, ksc495q7b, 2-bromomethyl-1,3 dioxolane, 2-bromomethyl-1, 3-dioxolane
78068
2-(bromomethyl)-1,3-dioxolane
C1COC(O1)CBr
4360-63-8
C4H7BrO2
167.002
MFCD00003214
CKIIJIDEWWXQEA-UHFFFAOYSA-N
2-bromomethyl-1,3-dioxolane, bromoacetaldehyde ethylene acetal, 1,3-dioxolane, 2-bromomethyl, 1,3-dioxolan-2-yl methyl bromide, bromomethyl dioxolane, acmc-1afrf, 2-bromomethyl-1,3-dioxolan, ksc495q7b, 2-bromomethyl-1,3 dioxolane, 2-bromomethyl-1, 3-dioxolane
78068
2-(bromomethyl)-1,3-dioxolane
C1COC(O1)CBr
2-Bromomethyl-1,3-dioxolane is used in the preparation of alfa, beta-unsaturated aldehyde and 1, 4-two aldehyde monoacetals synthesis of monomers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromomethyl-1,3-dioxolane is used in the preparation of alfa, beta-unsaturated aldehyde and 1, 4-two aldehyde monoacetals synthesis of monomers.
Solubility
Immiscible with water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY
Density 1.628
Boiling Point 175°C
Flash Point 62°C (145°F)
Refractive Index 1.482
Quantity 25 g
Beilstein 103309
Sensitivity Moisture sensitive
Solubility Information Immiscible with water.
Formula Weight 167.01
Percent Purity 97%
Chemical Name or Material 2-Bromomethyl-1,3-dioxolane