missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Bromomethyl)benzoic acid, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
2-(Bromomethyl)benzoic acid contains bi functional groups such as benzylic bromide and carboxylic acid are involved in nucleophilic reactions and esterification reactions respectively.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(Bromomethyl)benzoic acid contains bi functional groups such as benzylic bromide and carboxylic acid are involved in nucleophilic reactions and esterification reactions respectively.
Solubility
Insoluble in water.
Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.
2-(Bromomethyl)benzoic acid contains bi functional groups such as benzylic bromide and carboxylic acid are involved in nucleophilic reactions and esterification reactions respectively.
Solubility
Insoluble in water.
Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.
Specifications
Melting Point | ∽145°C (decomposition) |
Quantity | 10 g |
UN Number | UN3261 |
Solubility Information | Insoluble in water. |
Formula Weight | 215.05 |
Percent Purity | 97% |
Chemical Name or Material | 2-(Bromomethyl)benzoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'