missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-76486
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
18698-97-0
C8H7BrO2
215.05
MFCD00004314
DWXSYDKEWORWBT-UHFFFAOYSA-N
2-bromophenylacetic acid, 2-2-bromophenyl acetic acid, o-bromophenylacetic acid, 2-bromophenyl acetic acid, benzeneacetic acid, 2-bromo, 2-bromophenylaceticacid, 2-bromo-phenyl-acetic acid, pubchem7214, acmc-1btqg
87754
2-(2-bromophenyl)acetic acid
C1=CC=C(C(=C1)CC(=O)O)Br
This item is not returnable. View return policy
18698-97-0
C8H7BrO2
215.05
MFCD00004314
DWXSYDKEWORWBT-UHFFFAOYSA-N
2-bromophenylacetic acid, 2-2-bromophenyl acetic acid, o-bromophenylacetic acid, 2-bromophenyl acetic acid, benzeneacetic acid, 2-bromo, 2-bromophenylaceticacid, 2-bromo-phenyl-acetic acid, pubchem7214, acmc-1btqg
87754
2-(2-bromophenyl)acetic acid
C1=CC=C(C(=C1)CC(=O)O)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 104°C to 106°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula BrC6H4CH2COOH
Quantity 5 g
Formula Weight 215.05
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 2-Bromophenylacetic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromophenylacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'