missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothiophene, 98%, Thermo Scientific Chemicals

Catalog Number p-4510269
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
1003-09-4
C4H3BrS
163.03
MFCD00005417
TUCRZHGAIRVWTI-UHFFFAOYSA-N
2-thienyl bromide, thiophene, 2-bromo, thiophene, bromo, bromothiophene, 2-bromo-thiophene, 2-bromo thiophene, thienyl bromide, 5-bromothiophene, 5-bromo-thiophene, 2-bromanylthiophene
13851
2-bromothiophene
BrC1=CC=CS1
This item is not returnable. View return policy
1003-09-4
C4H3BrS
163.03
MFCD00005417
TUCRZHGAIRVWTI-UHFFFAOYSA-N
2-thienyl bromide, thiophene, 2-bromo, thiophene, bromo, bromothiophene, 2-bromo-thiophene, 2-bromo thiophene, thienyl bromide, 5-bromothiophene, 5-bromo-thiophene, 2-bromanylthiophene
13851
2-bromothiophene
BrC1=CC=CS1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -10.0°C
Color Brown to Colorless
Density 1.6800g/mL
Boiling Point 149.0°C to 151.0°C
Flash Point 52°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5850 to 1.5870
Quantity 50 mL
Beilstein 17,33
Specific Gravity 1.68
Solubility Information Solubility in water: immiscible. Other solubilities: very soluble in ether, acetone, soluble in carbontetrachloride
Formula Weight 163.04
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Bromothiophene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Bromothiophene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
Hazard Statement
  • H300-Fatal if swallowed.
  • H318-Causes serious eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'