Läs mer
2-C-Methyl-D-ribono-1,4-lactone, TRC
Produktkod. 30466491
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30466491 10 g
1 options
Produktkod. 30466491 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32596110G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-C-Methyl-D-ribonic-1,4-lactone
CAS 492-30-8
Molekylformel C6 H10 O5
Kvantitet 10 g
Synonym D-Ribonic acid, 2-C-methyl-, γ-lactone, Ribonic acid, 2-C-methyl-, γ-lactone, D- (7CI,8CI), 2-C-Methyl-D-ribo-pentonic acid γ-lactone, 2-C-Methyl-D-ribono-1,4-lactone, NSC 19768, NSC 62382, α-D-Glucosaccharinic acid γ-lactone
InChI formel InChI=1S/C6H10O5/c1-6(10)4(8)3(2-7)11-5(6)9/h3-4,7-8,10H,2H2,1H3/t3-,4-,6-/m1/s1
LEDER C[C@@]1(O)[C@H](O)[C@@H](CO)OC1=O
IUPAC-namn (3R,4R,5R)-3,4-dihydroxy-5-(hydroxymethyl)-3-methyloxolan-2-one
Molekylvikt (g/mol) 162.14
Formel vikt 162.0528