missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Carbamoyl-4-fluorobenzeneboronic acid pinacol ester, 96%

Brand:  Alfa Aesar™ H62909.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15477986

  • 5158.20 SEK / 1g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

2-Carbamoyl-4-fluorobenzeneboronic acid pinacol ester
265.09
2-Carbamoyl-4-fluorophenylboronic acid pinacol ester

Specifications

2-Carbamoyl-4-fluorobenzeneboronic acid pinacol ester
Solid
265.09
1g
96%
2-Carbamoyl-4-fluorophenylboronic acid pinacol ester
Videos
SDS