missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
2-Carboxy-N-methylpyridinium Iodide, Mikromol™
Beskrivning
2-Carboxy-N-methylpyridinium Iodide
Specifikationer
Specifikationer
| Kemiskt namn eller material | 2-Carboxy-N-methylpyridinium Iodide |
| CAS | 4-4-3785 |
| Analyt- eller komponentnamn | 2-Carboxy-N-methylpyridinium Iodide |
| Frakt skick | Room Temperature |
| Molekylformel | C7 H8 N O2 . I |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | 2-Carboxy-N-methylpyridinium Iodide |
| InChI formel | InChI=1S/C7H7NO2.HI/c1-8-5-3-2-4-6(8)7(9)10;/h2-5H,1H3;1H |
| LEDER | [I-].C[n+]1ccccc1C(=O)O |
| IUPAC-namn | 1-methylpyridin-1-ium-2-carboxylic acid;iodide |
| Visa mer |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?