Läs mer
2-(Carboxymethylamino) Nicotinic Acid Diethyl Ester, TRC
Produktkod. 30353104
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30353104 250 mg
1 options
Produktkod. 30353104 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC178738250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(Carboxymethylamino) Nicotinic Acid Diethyl Ester
CAS 1178176-00-5
Molekylformel C12H16N2O4
Kvantitet 250 mg
Synonym 2-[(2-Ethoxy-2-oxoethyl)amino]-3-pyridinecarboxylic Acid Ethyl Ester, Ethyl 2-(Ethylcarboxymethylamino)nicotinate
InChI formel InChI=1S/C12H16N2O4/c1-3-17-10(15)8-14-11-9(6-5-7-13-11)12(16)18-4-2/h5-7H,3-4,8H2,1-2H3,(H,13,14)
LEDER CCOC(=O)CNc1ncccc1C(=O)OCC
IUPAC-namn ethyl 2-[(2-ethoxy-2-oxoethyl)amino]pyridine-3-carboxylate
Molekylvikt (g/mol) 252.27
Formel vikt 252.111