Läs mer
∆2-Cefpodoxime Proxetil, TRC
Produktkod. 30134975
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134975 100 mg
30003954 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134975 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC243865100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 339528-86-8
Molekylformel C21H27N5O9S2
Molekylvikt (g/mol) 557.6
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-3-ene-2-carboxylic acid, 7-[[(2Z)-2-(2-amino-4-thiazolyl)-2-(methoxyimino)acetyl]amino]-3-(methoxymethyl)-8-oxo-, 1-[[(1-methylethoxy)carbonyl]oxy]ethyl ester, (6R,7R)-, 5-Thia-1-azabicyclo[4.2.0]oct-3-ene-2-carboxylic acid, 7-[[(2Z)-(2-amino-4-thiazolyl)(methoxyimino)acetyl]amino]-3-(methoxymethyl)-8-oxo-, 1-[[(1-methylethoxy)carbonyl]oxy]ethyl ester, (6R,7R)- (9CI), 1-[[(1-Methylethoxy)carbonyl]oxy]ethyl (6R,7R)-7-[[(2Z)-2-(2-amino-4-thiazolyl)-2-(methoxyimino)acetyl]amino]-3-(methoxymethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-3-ene-2-carboxylate, (1RS)-1-[[(1-Methylethoxy)carbonyl]oxy]ethyl (6R,7R)-7-[[(2Z)-2-(2-aminothiazol-4-yl)-2-(methoxyimino)acetyl]amino]-3-(methoxymethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-3-ene-2-carboxylate, Delta-2-cefpodoxime proxetil, Cefpodoxime proxetil Imp. C (EP)
IUPAC-namn 1-isopropoxycarbonyloxyethyl (6R,7R)-7-[[(2Z)-2-(2-aminothiazol-4-yl)-2-methoxyimino-acetyl]amino]-3-(methoxymethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-3-ene-2-carboxylate
LEDER COCC1=CS[C@@H]2[C@H](NC(=O)\C(=N/OC)\c3csc(N)n3)C(=O)N2C1C(=O)OC(C)OC(=O)OC(C)C
Specifikationer
Kemiskt namn eller material (Delta)2-Cefpodoxime Proxetil
Kvantitet 100 mg
InChI formel InChI=1S/C21H27N5O9S2/c1-9(2)33-21(30)35-10(3)34-19(29)15-11(6-31-4)7-36-18-14(17(28)26(15)18)24-16(27)13(25-32-5)12-8-37-20(22)23-12/h7-10,14-15,18H,6H2,1-5H3,(H2,22,23)(H,24,27)/b25-13-/t10?,14-,15?,18-/m1/s1
Formel vikt 557.125