Läs mer
2-Chloro-1,1,1-trimethoxyethane, TRC
Produktkod. 30360661
Change view
Kvantitet:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30360661 1 g
30354967 5 g
30349516 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30360661 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC4217801G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 74974-54-2
Molekylformel C5H11ClO3
Molekylvikt (g/mol) 154.59
Synonym 1-Chloro-2,2,2-trimethoxyethane, Trimethyl (Chloromethyl)orthoformate, Trimethyl Orthochloroacetate
IUPAC-namn 2-chloro-1,1,1-trimethoxyethane
LEDER ClCC(OC)(OC)OC
Specifikationer
Kemiskt namn eller material 2-Chloro-1,1,1-trimethoxyethane
Kvantitet 1 g
InChI formel InChI=1S/C5H11ClO3/c1-7-5(4-6,8-2)9-3/h4H2,1-3H3
Formel vikt 154.0397