Läs mer
2-Chloro-1,3-difluoro-4-nitrobenzene, 97%, Thermo Scientific™
Produktkod. 15476667
Change view
Kvantitet:
1 g
5 g
25 g
Förpackningsstorlek:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
15476667 1 g 1g
15486667 5 g 5g
15496667 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 15476667 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H33806.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3847-58-3
Molekylformel C6H2ClF2NO2
Molekylvikt (g/mol) 193.534
MDL-nummer MFCD00042201
InChI-nyckel CTVBVNKEMCGZDF-UHFFFAOYSA-N
Synonym 3-chloro-2,4-difluoronitrobenzene, 2,4-difluoro-3-chloronitrobenzene, benzene, 2-chloro-1,3-difluoro-4-nitro, 2,4-difluoro-3-chloronitrobnzene, 3-chloro-2,4-difluoro-1-nitrobenzene, 2-chloro-1,3-difluoro-4-nitro-benzene, pubchem4320, acmc-209j0b, ksc911k8t, buttpark 177\01-33
PubChem CID 223080
IUPAC-namn 2-chloro-1,3-difluoro-4-nitrobenzene
LEDER C1=CC(=C(C(=C1[N+](=O)[O-])F)Cl)F
Specifikationer
Kemiskt namn eller material 2-Chloro-1,3-difluoro-4-nitrobenzene
Smältpunkt 45°C to 49°C
Kvantitet 1 g
UN-nummer UN2923
Formel vikt 193.54
Procent renhet 97%