missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,3-thiazole-5-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-4391265
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
101012-12-8
C4H2ClNO2S
163.575
HNJOKQPEJIWTRF-UHFFFAOYSA-N
2-chlorothiazole-5-carboxylic acid, 2-chloro-5-thiazolecarboxylic acid, 5-thiazolecarboxylicacid, 2-chloro, 5-thiazolecarboxylic acid, 2-chloro, acmc-1brcy, 2-chlorothiazol-5-carboxylic acid, 2-chloro-5-carboxy-1,3-thiazole, 5-thiazolecarboxylicacid,2-chloro, 2-chloro-thiazole-5-carboxylic acid
1481389
2-chloro-1,3-thiazole-5-carboxylic acid
C1=C(SC(=N1)Cl)C(=O)O
This item is not returnable. View return policy
101012-12-8
C4H2ClNO2S
163.575
HNJOKQPEJIWTRF-UHFFFAOYSA-N
2-chlorothiazole-5-carboxylic acid, 2-chloro-5-thiazolecarboxylic acid, 5-thiazolecarboxylicacid, 2-chloro, 5-thiazolecarboxylic acid, 2-chloro, acmc-1brcy, 2-chlorothiazol-5-carboxylic acid, 2-chloro-5-carboxy-1,3-thiazole, 5-thiazolecarboxylicacid,2-chloro, 2-chloro-thiazole-5-carboxylic acid
1481389
2-chloro-1,3-thiazole-5-carboxylic acid
C1=C(SC(=N1)Cl)C(=O)O
Assay Percent Range 97%
Quantity 1 g
Formula Weight 163.58
Percent Purity 97%
Chemical Name or Material 2-Chloro-1, 3-thiazole-5-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'