Läs mer
2-Chloro-1-methylquinolinium Tetrafluoroborate, TRC
Produktkod. 30353591
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30353591 10 mg
30352357 50 mg
30349893 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30353591 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC60229510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 16843-24-6
Molekylformel C10H9ClN . BF4
Molekylvikt (g/mol) 265.44
Synonym Quinolinium, 2-chloro-1-methyl-, tetrafluoroborate(1-) (1:1), Quinolinium, 2-chloro-1-methyl-, tetrafluoroborate(1-) (8CI,9CI), Borate(1-), tetrafluoro-, 2-chloro-1-methylquinolinium (8CI), 1-Methyl-2-chloro-quinolinium tetrafluoroborate, 2-Chloro-1-methylquinolinium tetrafluoroborate
IUPAC-namn 2-chloro-1-methylquinolin-1-ium;tetrafluoroborate
LEDER C[n+]1c(Cl)ccc2ccccc12.F[B-](F)(F)F
Specifikationer
Kemiskt namn eller material 2-Chloro-1-methylquinolinium Tetrafluoroborate
Kvantitet 10 mg
InChI formel InChI=1S/C10H9ClN.BF4/c1-12-9-5-3-2-4-8(9)6-7-10(12)11;2-1(3,4)5/h2-7H,1H3;/q+1;-1
Formel vikt 265.0453