missing translation for 'onlineSavingsMsg'
Läs mer

2-Chloro-2-methylhexane, 97%

Produktkod. 11479616
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 g
25 g
Förpackningsstorlek:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11479616 5 g 5g
11489616 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11479616 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer B20183.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

CAS: 4398-65-6 | C7H15Cl | 134.65 g/mol

2-Chloro-2-methylhexane is used to produce other chemicals. For example, it is used to produce (1, 1-Dimethyl-pentyl)-benzene. Also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-2-methylhexane is used to produce other chemicals. For example, it is used to produce (1, 1-Dimethyl-pentyl)-benzene. Also used as a pharmaceutical intermediate.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 4398-65-6
Molekylformel C7H15Cl
Molekylvikt (g/mol) 134.65
MDL-nummer MFCD00060779
InChI-nyckel KBOBQLJBYKKAPN-UHFFFAOYSA-N
Synonym hexane, 2-chloro-2-methyl, acmc-1an7f, 2-chloro-2-methyl-hexane, 1,1-dimethylpentyl chloride, 1-chloro-1,1-dimethylpentane, 2-chloro-2-methylhexane 5g
PubChem CID 138224
IUPAC-namn 2-chloro-2-methylhexane
LEDER CCCCC(C)(C)Cl

Specifikationer

Kemiskt namn eller material 2-Chloro-2-methylhexane
Kvantitet 5 g
UN-nummer UN1993
Formel vikt 134.65
Procent renhet 97%

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.