missing translation for 'onlineSavingsMsg'
Läs mer

2-Chloro-2-methylpropane, Sigma-Aldrich™

Produktkod. 15858580
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mL
1 L
Förpackningsstorlek:
1 L
100ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
15858580 1 L 1 L
15848580 100 mL 100ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15858580 Leverantör Sigma Aldrich Leverantörsnummer 8015501000
 Begränsad artikel
Beräknad leverans: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 507-20-0
Kvalitet Research
Molekylvikt (g/mol) 92.57

Specifications

Smältpunkt -28°C
Densitet 0.84 g/cm3 at 20°C
MDL-nummer MFCD00000816
Kvantitet 1 L
Synonym 2-Chloro-2-methylpropane, tert-Butyl chloride, 2-Chloro-2-methylpropane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.