Learn More
2-Chloro-3,5-bis(trifluoromethyl)aniline, 98%, BLD Pharm™
Product Code. 31070978
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31070978 500 g
31064144 5 g
31062631 100 g
31058522 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 31070978 Supplier BLD Pharm Supplier No. BD10825500g
 Restricted Item
Beräknad leverans: 27-11-2026
Specifications
CAS 201593-90-0
Molecular Formula C8H4ClF6N
MDL Number MFCD04038716
Quantity 500 g
SMILES NC1=C(Cl)C(=CC(=C1)C(F)(F)F)C(F)(F)F
Molecular Weight (g/mol) 263.57
Percent Purity 98%
Chemical Name or Material 2-Chloro-3,5-bis(trifluoromethyl)aniline