missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3,5-bis(trifluoromethyl)aniline, 98%, Thermo Scientific™

Catalog Number 15491169
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
201593-90-0
C8H4ClF6N
263.567
MFCD04038716
JKUFETFGEJPHEM-UHFFFAOYSA-N
2-chloro-3,5-bis trifluoromethyl aniline, 3,5-bis trifluoromethyl-2-chloroaniline, 2-chloro-3,5-bis-trifluoromethyl aniline, benzenamine, 2-chloro-3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl-2-chlorophenylamine, pubchem2938, 2-chloro-3,5-bis-trifluoromethyl-aniline, 3,5-bis trifluoromethyl-2-chloro aniline, 2-chloranyl-3,5-bis trifluoromethyl aniline, 2-chloro-3,5-bis-trifluoromethyl-phenylamine
2757429
2-chloro-3,5-bis(trifluoromethyl)aniline
C1=C(C=C(C(=C1C(F)(F)F)Cl)N)C(F)(F)F
This item is not returnable. View return policy
201593-90-0
C8H4ClF6N
263.567
MFCD04038716
JKUFETFGEJPHEM-UHFFFAOYSA-N
2-chloro-3,5-bis trifluoromethyl aniline, 3,5-bis trifluoromethyl-2-chloroaniline, 2-chloro-3,5-bis-trifluoromethyl aniline, benzenamine, 2-chloro-3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl-2-chlorophenylamine, pubchem2938, 2-chloro-3,5-bis-trifluoromethyl-aniline, 3,5-bis trifluoromethyl-2-chloro aniline, 2-chloranyl-3,5-bis trifluoromethyl aniline, 2-chloro-3,5-bis-trifluoromethyl-phenylamine
2757429
2-chloro-3,5-bis(trifluoromethyl)aniline
C1=C(C=C(C(=C1C(F)(F)F)Cl)N)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Chloro-3,5-bis(trifluoromethyl)aniline is used as dyestuff Intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.595 g/mL
Refractive Index 1.459
Quantity 5 g
UN Number UN2810
Beilstein 13730810
Solubility Information Slightly soluble in water.
Formula Weight 263.57
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Chloro-3,5-bis(trifluoromethyl)aniline
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'