missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-nitropyridine, 99+%, Thermo Scientific Chemicals

Catalog Number p-3777920
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5470-18-8
C5H3ClN2O2
158.54
MFCD00006232
UUOLETYDNTVQDY-UHFFFAOYSA-N
pyridine, 2-chloro-3-nitro, 3-nitro-2-chloropyridine, 2-chloro-3-nitro-pyridine, 2-chloro-3-nitro pyridine, 2-chlor-3-nitropyridin, zlchem 303, pubchem1229, 2-chloro-nitropyridine, 2-chloro-3nitropyridine, 2-chloro3-nitropyridine
79613
2-chloro-3-nitropyridine
C1=CC(=C(N=C1)Cl)[N+](=O)[O-]
This item is not returnable. View return policy
5470-18-8
C5H3ClN2O2
158.54
MFCD00006232
UUOLETYDNTVQDY-UHFFFAOYSA-N
pyridine, 2-chloro-3-nitro, 3-nitro-2-chloropyridine, 2-chloro-3-nitro-pyridine, 2-chloro-3-nitro pyridine, 2-chlor-3-nitropyridin, zlchem 303, pubchem1229, 2-chloro-nitropyridine, 2-chloro-3nitropyridine, 2-chloro3-nitropyridine
79613
2-chloro-3-nitropyridine
C1=CC(=C(N=C1)Cl)[N+](=O)[O-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point 100.0°C to 103.0°C
Color Yellow
Flash Point 185°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Linear Formula Cl(C5H3N)NO2
Refractive Index 1.4540 to 1.4560
Quantity 25 g
Beilstein 20, I, 82
Solubility Information Solubility in water: insoluble
Formula Weight 158.54
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 2-chloro-3-nitropyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-3-nitropyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'