missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-(trifluoromethyl)benzyl alcohol, 97+%, Thermo Scientific™

Catalog Number p-7026905
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
261763-20-6
C8H6ClF3O
210.58
MFCD01631537
MQUXXVLMXCHGDZ-UHFFFAOYSA-N
2-chloro-3-trifluoromethyl benzyl alcohol, 2-chloro-3-trifluoromethyl phenyl methanol, 2-chloro-3-trifluoromethyl benzylalcohol, 2-chloro-3-trifluoromethyl phenyl methan-1-ol, acmc-1coey, rarechem al bd 0475, rarechem al bd 1301, 2-chloro-3-trifluoromethyl-benzylalcohol, 2-chloro-3-trifluoromethylbenzyl alcohol
2778108
[2-chloro-3-(trifluoromethyl)phenyl]methanol
C1=CC(=C(C(=C1)C(F)(F)F)Cl)CO
This item is not returnable. View return policy
261763-20-6
C8H6ClF3O
210.58
MFCD01631537
MQUXXVLMXCHGDZ-UHFFFAOYSA-N
2-chloro-3-trifluoromethyl benzyl alcohol, 2-chloro-3-trifluoromethyl phenyl methanol, 2-chloro-3-trifluoromethyl benzylalcohol, 2-chloro-3-trifluoromethyl phenyl methan-1-ol, acmc-1coey, rarechem al bd 0475, rarechem al bd 1301, 2-chloro-3-trifluoromethyl-benzylalcohol, 2-chloro-3-trifluoromethylbenzyl alcohol
2778108
[2-chloro-3-(trifluoromethyl)phenyl]methanol
C1=CC(=C(C(=C1)C(F)(F)F)Cl)CO

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 74°C to 77°C
Quantity 1 g
Formula Weight 210.58
Percent Purity ≥97%
Chemical Name or Material 2-Chloro-3-(trifluoromethyl)benzyl alcohol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'