missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-bromo-5-fluorobenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number p-7055616
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1214386-29-4
C7H3BrClFO
237.452
MFCD13185877
XPEGZWLIWARWAA-UHFFFAOYSA-N
2-chloro-4-bromo-5-fluorobenzaldehyde
70680695
4-bromo-2-chloro-5-fluorobenzaldehyde
C1=C(C(=CC(=C1F)Br)Cl)C=O
This item is not returnable. View return policy
1214386-29-4
C7H3BrClFO
237.452
MFCD13185877
XPEGZWLIWARWAA-UHFFFAOYSA-N
2-chloro-4-bromo-5-fluorobenzaldehyde
70680695
4-bromo-2-chloro-5-fluorobenzaldehyde
C1=C(C(=CC(=C1F)Br)Cl)C=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Formula Weight 237.45
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Chloro-4-bromo-5-fluorobenzaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'