Läs mer
2-Chloro-4-[(dimethylcarbamoyl)amino]benzoic Acid, TRC
Produktkod. 30451044
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30451044 100 mg
1 options
Produktkod. 30451044 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT776845100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Chlorotoluron-benzoic Acid
CAS 59587-01-8
Molekylformel C10 H11 Cl N2 O3
Kvantitet 100 mg
Synonym Chlorotoluron-benzoic Acid, 2-Chloro-4-[[(dimethylamino)carbonyl]amino]benzoic acid (ACI), 2-Chloro-4-[(dimethylcarbamoyl)amino]benzoic acid, CGA 15140, N-(3-Chloro-4-carboxyphenyl)-N',N'-dimethylurea, Benzoic acid','2-chloro-4-[[(dimethylamino)carbonyl]amino]- (9CI','ACI)
InChI formel InChI=1S/C10H11ClN2O3/c1-13(2)10(16)12-6-3-4-7(9(14)15)8(11)5-6/h3-5H,1-2H3,(H,12,16)(H,14,15)
LEDER CN(C)C(=O)Nc1ccc(C(=O)O)c(Cl)c1
IUPAC-namn 2-chloro-4-(dimethylcarbamoylamino)benzoic acid
Molekylvikt (g/mol) 242.6589
Formel vikt 242.0458