Läs mer
2-Chloro-4-hydroxy-5-methoxybenzaldehyde, TRC
Produktkod. 30354850
Change view
Kvantitet:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30354850 50 mg
30352185 25 mg
30354740 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30354850 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC61162050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 18268-76-3
Molekylformel C8 H7 Cl O3
Molekylvikt (g/mol) 186.59
Synonym 6-Chlorovanillin, Benzaldehyde, 2-chloro-4-hydroxy-5-methoxy-, Vanillin, 6-chloro- (8CI), 2-Chloro-4-hydroxy-5-methoxybenzaldehyde, 6-Chlorovanillin, 6-Monochlorovanillin, NSC 95796
IUPAC-namn 2-chloro-4-hydroxy-5-methoxybenzaldehyde
LEDER COc1cc(C=O)c(Cl)cc1O
Specifikationer
Kemiskt namn eller material 6-Chlorovanillin
Kvantitet 50 mg
InChI formel InChI=1S/C8H7ClO3/c1-12-8-2-5(4-10)6(9)3-7(8)11/h2-4,11H,1H3
Formel vikt 186.0084