missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methylbenzotrifluoride, 97+%, Thermo Scientific™

Catalog Number p-7027037
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
74483-46-8
C8H6ClF3
194.581
MFCD00190112
OHDYNLHQHOFWGR-UHFFFAOYSA-N
2-chloro-4-methylbenzotrifluoride, 2-chloro-4-methyl-1-trifluoromethyl benzene, 3-chloro-4-trifluoromethyl toluene, 2-chloro-4-methyl-1-trifluoromethyl-benzene, benzene, 2-chloro-4-methyl-1-trifluoromethyl, pubchem4851, ksc954c8j, 2-chloro4-methyl-1-trifluoromethylbenzene
2736619
2-chloro-4-methyl-1-(trifluoromethyl)benzene
CC1=CC(=C(C=C1)C(F)(F)F)Cl
This item is not returnable. View return policy
74483-46-8
C8H6ClF3
194.581
MFCD00190112
OHDYNLHQHOFWGR-UHFFFAOYSA-N
2-chloro-4-methylbenzotrifluoride, 2-chloro-4-methyl-1-trifluoromethyl benzene, 3-chloro-4-trifluoromethyl toluene, 2-chloro-4-methyl-1-trifluoromethyl-benzene, benzene, 2-chloro-4-methyl-1-trifluoromethyl, pubchem4851, ksc954c8j, 2-chloro4-methyl-1-trifluoromethylbenzene
2736619
2-chloro-4-methyl-1-(trifluoromethyl)benzene
CC1=CC(=C(C=C1)C(F)(F)F)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 194.58
Percent Purity ≥97%
Chemical Name or Material 2-Chloro-4-methylbenzotrifluoride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'