Läs mer
2-Chloro-4-nitrophenyl Alpha-D-Maltotrioside, TRC
Produktkod. 30136017
Change view
Kvantitet:
25 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136017 25 mg
30137492 500 mg
17928304 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136017 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC59647325MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 118291-90-0
Molekylformel C24H34ClNO18
Molekylvikt (g/mol) 659.98
Synonym 2-Chloro-4-nitrophenyl alpha-Maltotrioside, 2-Chloro-4-nitrophenyl O-alpha-D-glucopyranosyl-(1-4)-O-alpha-D-glucopyranosyl-(1-4)-alpha-D-glucopyranoside
IUPAC-namn (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-[(2R,3S,4R,5R,6R)-6-(2-chloro-4-nitrophenoxy)-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
LEDER OC[C@H]1OC(O[C@H]2C(O)[C@@H](O)C(O[C@H]3C(O)[C@@H](O)C(Oc4ccc(cc4Cl)[N+](=O)[O-])O[C@@H]3CO)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
Specifikationer
Kemiskt namn eller material 2-Chloro-4-nitrophenyl alpha-D-Maltotrioside
Kvantitet 25 mg
InChI formel InChI=1S/C24H34ClNO18/c25-8-3-7(26(37)38)1-2-9(8)39-22-18(35)15(32)20(11(5-28)41-22)44-24-19(36)16(33)21(12(6-29)42-24)43-23-17(34)14(31)13(30)10(4-27)40-23/h1-3,10-24,27-36H,4-6H2/t10-,11-,12-,13-,14+,15-,16-,17-,18-,19-,20-,21-,22+,23-,24-/m1/s1
Formel vikt 659.1464