missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-nitropyridine, 98%, ACROS Organics™

Brand:  Acros Organics 443530050

Additional Details : CAS Number : 23056-36-2 Weight : 0.01000kg


Quantity 5g

 View more versions of this product

Product Code. 15239626

  • 1701.00 SEK / 5g

Estimated Shipment: 08-07-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

23056-36-2
158.54
LIEPVGBDUYKPLC-UHFFFAOYSA-N
735152
[O-][N+](=O)C1=CC(Cl)=NC=C1
C5H3ClN2O2
MFCD00661454
pyridine, 2-chloro-4-nitro, 2-chloro-4-nitro-pyridine, 2-chlor-4-nitropyridin, pubchem9220, 2-chloro-4nitropyridine, 2chloro-4-nitropyridine, acmc-1ccv7, 2-chloro-4-nitro pyridine, ksc201q1f, 2-chloro-4-nitropyridine
2-chloro-4-nitropyridine

Specifications

2-Chloro-4-nitropyridine
5g
97.5% min. (GC)
MFCD00661454
LIEPVGBDUYKPLC-UHFFFAOYSA-N
2-chloro-4-nitropyridine
735152
98%
52.0°C to 56.0°C
23056-36-2
C5H3ClN2O2
pyridine, 2-chloro-4-nitro, 2-chloro-4-nitro-pyridine, 2-chlor-4-nitropyridin, pubchem9220, 2-chloro-4nitropyridine, 2chloro-4-nitropyridine, acmc-1ccv7, 2-chloro-4-nitro pyridine, ksc201q1f, 2-chloro-4-nitropyridine
[O-][N+](=O)C1=CC(Cl)=NC=C1
158.54
158.54
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Chloro-4-nitropyridine, 98%

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents
Certificates