Läs mer
2-Chloro-4-(tert-pentyl)phenol, TRC
Produktkod. 30180071
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180071 50 mg
30179900 500 mg
30180262 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30180071 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC42179350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5323-65-9
Molekylformel C11H15OCl
Molekylvikt (g/mol) 198.68
IUPAC-namn 2-chloro-4-(2-methylbutan-2-yl)phenol
LEDER CCC(C)(C)c1ccc(O)c(Cl)c1
Specifikationer
Kemiskt namn eller material 2-Chloro-4-(tert-pentyl)phenol
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C11H15ClO/c1-4-11(2,3)8-5-6-10(13)9(12)7-8/h5-7,13H,4H2,1-3H3
Formel vikt 198.0811
Procent renhet >95%