missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-4-thien-2-ylpyrimidine, Thermo Scientific™, 97%, Thermo Scientific™

Catalog Number p-3175019
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
83726-75-4
C8H5ClN2S
196.652
MFCD08690327
DTDHCBGOLGIIOA-UHFFFAOYSA-N
14117941
2-chloro-4-thiophen-2-ylpyrimidine
C1=CSC(=C1)C2=NC(=NC=C2)Cl
This item is not returnable. View return policy
83726-75-4
C8H5ClN2S
196.652
MFCD08690327
DTDHCBGOLGIIOA-UHFFFAOYSA-N
14117941
2-chloro-4-thiophen-2-ylpyrimidine
C1=CSC(=C1)C2=NC(=NC=C2)Cl
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 2-chloro-4-thiophen-2-yl pyrimidine, 2-chloro-4-thien-2-ylpyrimidine, pyrimidine, 2-chloro-4-2-thienyl, 2-chloro-4-thiophen-2-yl-pyrimidine, 2-chloro-4-2-thienyl pyrimidine, 2-chloro-4-thien-2-yl pyrimidine, 2-2-chloropyrimidin-4-yl thiophene, pyrimidine,2-chloro-4-2-thienyl
Formula Weight 196.66
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-thien-2-ylpyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'