missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(trifluoromethyl)pyridine, 95%, Thermo Scientific™

Catalog Number p-7077508
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
81565-18-6
C6H3ClF3N
181.54
MFCD00042224
GBNPVXZNWBWNEN-UHFFFAOYSA-N
2-chloro-4-trifluoromethyl pyridine, pyridine, 2-chloro-4-trifluoromethyl, 2-chloro-4-trifluoro methylpyridine, 2-chloro-4-trifluoromethyl-pyridine, pubchem2204, acmc-209plt, ksc449e5r, 2-chloro-4-trifluromethylpyridine, 2-chloro-4-trifluoro-methylpyridine
144958
2-chloro-4-(trifluoromethyl)pyridine
FC(F)(F)C1=CC=NC(Cl)=C1
This item is not returnable. View return policy
81565-18-6
C6H3ClF3N
181.54
MFCD00042224
GBNPVXZNWBWNEN-UHFFFAOYSA-N
2-chloro-4-trifluoromethyl pyridine, pyridine, 2-chloro-4-trifluoromethyl, 2-chloro-4-trifluoro methylpyridine, 2-chloro-4-trifluoromethyl-pyridine, pubchem2204, acmc-209plt, ksc449e5r, 2-chloro-4-trifluromethylpyridine, 2-chloro-4-trifluoro-methylpyridine
144958
2-chloro-4-(trifluoromethyl)pyridine
FC(F)(F)C1=CC=NC(Cl)=C1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.4110g/mL
Boiling Point 48.0°C (8.0 mmHg)
Flash Point 40°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 1.411
Formula Weight 181.54
Percent Purity 95%
Chemical Name or Material 2-Chloro-4-(trifluoromethyl)pyridine
Vedi altri risultati Mostra meno risultati
compliance-icons compliance-icons
Identificativo prodotto
  • 2-Chloro-4-(trifluoromethyl)pyridine, 95%
Avvertenza
  • Warning
Categoria di pericolo
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Consigli di prudenza
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'