Läs mer
2-Chloro-4-(trifluoromethyl)pyridine, TRC
Produktkod. 30354339
Change view
Kvantitet:
2.5 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30354339 5 g
30355194 2.5 g
30350795 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30354339 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC5974885G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 81565-18-6
Molekylformel C6H3ClF3N
Molekylvikt (g/mol) 181.54
IUPAC-namn 2-chloro-4-(trifluoromethyl)pyridine
LEDER FC(F)(F)c1ccnc(Cl)c1
Specifikationer
Kemiskt namn eller material 2-Chloro-4-(trifluoromethyl)pyridine
Kvantitet 5 g
InChI formel InChI=1S/C6H3ClF3N/c7-5-3-4(1-2-11-5)6(8,9)10/h1-3H
Formel vikt 180.9906