Läs mer
2-Chloro-5-(1-ethoxy-3-oxoisoindolin-1-yl)benzenesulfonamide, TRC
Produktkod. 30367408
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30367408 250 mg
30354104 100 mg
30351588 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30367408 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC367146250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1369995-36-7
Molekylformel C16H15ClN2O4S
Molekylvikt (g/mol) 366.82
Synonym Chlorthalidone Impurity
IUPAC-namn 2-chloro-5-(1-ethoxy-3-oxo-2H-isoindol-1-yl)benzenesulfonamide
LEDER CCOC1(NC(=O)c2ccccc12)c3ccc(Cl)c(c3)S(=O)(=O)N
Specifikationer
Kemiskt namn eller material 2-Chloro-5-(1-ethoxy-3-oxoisoindolin-1-yl)benzenesulfonamide
Kvantitet 250 mg
InChI formel InChI=1S/C16H15ClN2O4S/c1-2-23-16(12-6-4-3-5-11(12)15(20)19-16)10-7-8-13(17)14(9-10)24(18,21)22/h3-9H,2H2,1H3,(H,19,20)(H2,18,21,22)
Formel vikt 366.0441