Läs mer
2-Chloro-5,6-dimethyl-4-methoxy Nicotinic Acid Methyl Ester, TRC
Produktkod. 30356219
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30356219 250 mg
1 options
Produktkod. 30356219 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC365435250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Chloro-5,6-dimethyl-4-methoxy Nicotinic Acid Methyl Ester
CAS 1329833-74-0
Molekylformel C10H12ClNO3
Kvantitet 250 mg
Synonym 2-Chloro-4-methoxy-5,6-dimethyl-3-pyridinecarboxylic Acid Methyl Ester, Methyl 2-Chloro-4-methoxy-5,6-dimethyl-3-pyridinecarboxylate
InChI formel InChI=1S/C10H12ClNO3/c1-5-6(2)12-9(11)7(8(5)14-3)10(13)15-4/h1-4H3
LEDER COC(=O)c1c(Cl)nc(C)c(C)c1OC
IUPAC-namn methyl 2-chloro-4-methoxy-5,6-dimethylpyridine-3-carboxylate
Molekylvikt (g/mol) 229.66
Formel vikt 229.0506