Learn More
2-chloro-5-ethylpyrimidine, 97%, Thermo Scientific™
Catalog Number p-4258894
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
111196-81-7
C6H7ClN2
142.59
MFCD00799503
BGLLZQRUXJGTAD-UHFFFAOYSA-N
5-ethyl-2-chloropyrimidine, 2-chloro-5-ethyl-pyrimidine, pyrimidine, 2-chloro-5-ethyl, 2-chloro-5-ethyl pyrimidine, pubchem7032, acmc-1brw8, 2-chloro-5-ethylpyrimdine, 2-chloro-5-ethylpyrimidin, 2-chloro-5-ethylpyrimidine, ksc493s1b
3572763
2-chloro-5-ethylpyrimidine
CCC1=CN=C(Cl)N=C1
111196-81-7
C6H7ClN2
142.59
MFCD00799503
BGLLZQRUXJGTAD-UHFFFAOYSA-N
5-ethyl-2-chloropyrimidine, 2-chloro-5-ethyl-pyrimidine, pyrimidine, 2-chloro-5-ethyl, 2-chloro-5-ethyl pyrimidine, pubchem7032, acmc-1brw8, 2-chloro-5-ethylpyrimdine, 2-chloro-5-ethylpyrimidin, 2-chloro-5-ethylpyrimidine, ksc493s1b
3572763
2-chloro-5-ethylpyrimidine
CCC1=CN=C(Cl)N=C1
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1740g/mL
Boiling Point 223°C
Flash Point >109°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 1.174
Formula Weight 142.59
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 2-chloro-5-ethylpyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-5-ethylpyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.