Learn More
Alfa Aesar™ 2-Chloro-5-fluorobenzeneboronic acid pinacol ester, 96%
Brand:  Alfa Aesar™ H62628.MD
Additional Details : CAS Number : 870486-41-2 Weight : 0.00025kg
Quantity 250mg
 View more versions of this product
Product Code. 15497646
  • 2122.38 SEK / 250mg
Estimated Shipment date
from Supplier 25-07-2019
Add to basket
Specifications
Chemical Identifiers
870486-41-2
256.508
ZDEZMLTVDFZRET-UHFFFAOYSA-N
59218658
B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC(=C2)F)Cl
C12H15BClFO2
MFCD16996257
2-2-chloro-5-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-chloro-5-fluorophenylboronic acid pinacol ester, 2-chloro-5-fluorobenzeneboronic acid pinacol ester, 2-2-chloro-5-fluoro-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 1,3,2-dioxaborolane, 2-2-chloro-5-fluorophenyl-4,4,5,5-tetramethyl
2-(2-chloro-5-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Specifications
2-Chloro-5-fluorobenzeneboronic acid pinacol ester
870486-41-2
MFCD16996257
ZDEZMLTVDFZRET-UHFFFAOYSA-N
2-(2-chloro-5-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
59218658
Solid
250mg
C12H15BClFO2
2-2-chloro-5-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-chloro-5-fluorophenylboronic acid pinacol ester, 2-chloro-5-fluorobenzeneboronic acid pinacol ester, 2-2-chloro-5-fluoro-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 1,3,2-dioxaborolane, 2-2-chloro-5-fluorophenyl-4,4,5,5-tetramethyl
B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC(=C2)F)Cl
256.508
256.51
96%