missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-pyridineboronic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-6827173
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
444120-91-6
C5H5BClNO2
157.36
MFCD03094998
WPAPNCXMYWRTTL-UHFFFAOYSA-N
2-chloropyridine-5-boronic acid, 6-chloropyridin-3-yl boronic acid, 2-chloro-5-pyridineboronic acid, 2-chloro-5-pyridylboronic acid, 6-chloro-3-pyridylboronic acid, 6-chloropyridin-3-yl-3-boronic acid, 6-chloropyridine-3-boronic acid, 2-chloropyridin-5-ylboronic acid, 6-chloro-3-pyridinylboronic acid, 2-chloro-pyridine-5-boronic acid
3845034
(6-chloropyridin-3-yl)boronic acid
OB(O)C1=CC=C(Cl)N=C1
This item is not returnable. View return policy
444120-91-6
C5H5BClNO2
157.36
MFCD03094998
WPAPNCXMYWRTTL-UHFFFAOYSA-N
2-chloropyridine-5-boronic acid, 6-chloropyridin-3-yl boronic acid, 2-chloro-5-pyridineboronic acid, 2-chloro-5-pyridylboronic acid, 6-chloro-3-pyridylboronic acid, 6-chloropyridin-3-yl-3-boronic acid, 6-chloropyridine-3-boronic acid, 2-chloropyridin-5-ylboronic acid, 6-chloro-3-pyridinylboronic acid, 2-chloro-pyridine-5-boronic acid
3845034
(6-chloropyridin-3-yl)boronic acid
OB(O)C1=CC=C(Cl)N=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 165°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 157.36
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-pyridineboronic acid
compliance-icons
Product Identifier
  • 2-Chloro-5-pyridineboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'