missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylaniline, 98%, Thermo Scientific Chemicals

Catalog Number p-7043768
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
87-63-8
C7H8ClN
141.60
MFCD00007679
WFNLHDJJZSJARK-UHFFFAOYSA-N
6-chloro-2-methylaniline, 6-chloro-o-toluidine, 2-amino-3-chlorotoluene, benzenamine, 2-chloro-6-methyl, 3-chloro-2-aminotoluene, 6-chloro-2-toluidine, 2-methyl-6-chloroaniline, o-toluidine, 6-chloro, 2-chloro-6-methyl-aniline, 2-chloro-6-methylaninile
6897
2-chloro-6-methylaniline
CC1=CC=CC(Cl)=C1N
This item is not returnable. View return policy
87-63-8
C7H8ClN
141.60
MFCD00007679
WFNLHDJJZSJARK-UHFFFAOYSA-N
6-chloro-2-methylaniline, 6-chloro-o-toluidine, 2-amino-3-chlorotoluene, benzenamine, 2-chloro-6-methyl, 3-chloro-2-aminotoluene, 6-chloro-2-toluidine, 2-methyl-6-chloroaniline, o-toluidine, 6-chloro, 2-chloro-6-methyl-aniline, 2-chloro-6-methylaninile
6897
2-chloro-6-methylaniline
CC1=CC=CC(Cl)=C1N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 2°C
Density 1.174
Boiling Point 215°C
Flash Point 98°C (208°F)
Refractive Index 1.5775
Quantity 1 g
UN Number UN3429
Beilstein 774624
Formula Weight 141.6
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-methylaniline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'