Learn More
(2-Chloro-7H-purin-6-yl)-furan-2-ylmethyl-amine, 98%, BLD Pharm™
Product Code. 31033599
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
Unit Size:
100mg
1g
250mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31033599 1 g 1g
31025981 100 mg 100mg
31015612 5 g 5g
31024325 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 31033599 Supplier BLD Pharm Supplier No. BD008036751g
 Restricted Item
Beräknad leverans: 27-11-2026
Specifications
CAS 101862-47-9
Molecular Formula C10H8ClN5O
MDL Number MFCD09792860
Quantity 1 g
SMILES ClC1=NC(NCC2=CC=CO2)=C3N=CNC3=N1
Molecular Weight (g/mol) 249.66
Percent Purity 98%
Chemical Name or Material (2-Chloro-7H-purin-6-yl)-furan-2-ylmethyl-amine