missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-8-fluoroquinoline, 95%, Thermo Scientific™

Catalog Number 15446257
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
124467-23-8
C9H5ClFN
181.594
MFCD16140178
CSPMWYOOKWBXRI-UHFFFAOYSA-N
22125194
2-chloro-8-fluoroquinoline
C1=CC2=C(C(=C1)F)N=C(C=C2)Cl
This item is not returnable. View return policy
124467-23-8
C9H5ClFN
181.594
MFCD16140178
CSPMWYOOKWBXRI-UHFFFAOYSA-N
22125194
2-chloro-8-fluoroquinoline
C1=CC2=C(C(=C1)F)N=C(C=C2)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 82°C to 84°C
Quantity 250 mg
Synonym quinoline, 2-chloro-8-fluoro, acmc-209as5, quinoline,2-chloro-8-fluoro, 2-chloro-8-fluoroquinoline
Formula Weight 181.6
Percent Purity 95%
Chemical Name or Material 2-Chloro-8-fluoroquinoline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'