Läs mer
2-Chloro-9-fluorenone, TRC
Produktkod. 30366006
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30366006 50 mg
30361065 250 mg
30361599 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30366006 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36626350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3096-47-7
Molekylformel C13 H7 Cl O
Molekylvikt (g/mol) 214.65
Synonym 2-Chloro-9H-fluoren-9-one, 2-Chloro-9-fluorenone, 2-Chlorofluorenone, NSC 107566
IUPAC-namn 2-chlorofluoren-9-one
LEDER Clc1ccc2c3ccccc3C(=O)c2c1
Specifikationer
Kemiskt namn eller material 2-Chloro-9-fluorenone
Kvantitet 50 mg
InChI formel InChI=1S/C13H7ClO/c14-8-5-6-10-9-3-1-2-4-11(9)13(15)12(10)7-8/h1-7H
Formel vikt 214.0185