Läs mer
2-Chloro-DL-phenylalanine, TRC
Produktkod. 30363893
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30363893 1 g
1 options
Produktkod. 30363893 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3751451G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Chloro-DL-phenylalanine
CAS 14091-11-3
Molekylformel C9 H10 Cl N O2
Kvantitet 1 g
Synonym Phenylalanine, 2-chloro-, Alanine, 3-(o-chlorophenyl)-, DL- (8CI), DL-Phenylalanine, 2-chloro-, DL-2-Chlorophenylalanine, NSC 154946, NSC 233876, NSC 28185, o-Chloro-DL-phenylalanine
InChI formel InChI=1S/C9H10ClNO2/c10-7-4-2-1-3-6(7)5-8(11)9(12)13/h1-4,8H,5,11H2,(H,12,13)
LEDER NC(Cc1ccccc1Cl)C(=O)O
IUPAC-namn 2-amino-3-(2-chlorophenyl)propanoic acid
Molekylvikt (g/mol) 199.63
Formel vikt 199.04