Läs mer
2-Chloro Loratadine, TRC
Produktkod. 30349735
Change view
Kvantitet:
1 mg
2.5 mg
5 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30349735 5 mg -
30360627 1 mg -
30355655 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30349735 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3754705MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 165739-64-0
Molekylformel C22 H22 Cl2 N2 O2
Molekylvikt (g/mol) 417.33
Synonym Ethyl 4-(2,8-Dichloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)piperidine-1-carboxylate
IUPAC-namn ethyl 4-(2,8-dichloro-5,6-dihydrobenzo[1,2]cyclohepta[2,4-c]pyridin-11-ylidene)piperidine-1-carboxylate
LEDER CCOC(=O)N1CCC(=C2c3ccc(Cl)cc3CCc4ccc(Cl)nc24)CC1
Specifikationer
Kemiskt namn eller material Ethyl 4-(2,8-Dichloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)piperidine-1-carboxylate
Kvantitet 5 mg
InChI formel InChI=1S/C22H22Cl2N2O2/c1-2-28-22(27)26-11-9-14(10-12-26)20-18-7-6-17(23)13-16(18)4-3-15-5-8-19(24)25-21(15)20/h5-8,13H,2-4,9-12H2,1H3
Formel vikt 416.1058