Läs mer
2-Chloro-N-(2,6-dichlorophenyl)-N-phenylacetamide, Mikromol™
Produktkod. 17923406
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17923406 25 mg
17921136 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17923406 Leverantör Mikromol Leverantörsnummer MM0006.190025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Chloro-N-(2,6-dichlorophenyl)-N-phenylacetamide
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Chloro-N-(2,6-dichlorophenyl)-N-phenylacetamide
CAS 15308-01-7
Analyt- eller komponentnamn 2-Chloro-N-(2,6-dichlorophenyl)-N-phenylacetamide
Frakt skick Room Temperature
Molekylformel C14 H10 Cl3 N O
Certifikat och efterlevnad ISO 17025
Synonym 2-Chloro-N-(2,6-dichlorophenyl)-N-phenylacetamide
InChI formel InChI=1S/C14H10Cl3NO/c15-9-13(19)18(10-5-2-1-3-6-10)14-11(16)7-4-8-12(14)17/h1-8H,9H2
LEDER ClCC(=O)N(c1ccccc1)c2c(Cl)cccc2Cl
IUPAC-namn 2-chloro-N-(2,6-dichlorophenyl)-N-phenylacetamide
Molekylvikt (g/mol) 314.59
Kvantitet 25 mg
Formel vikt 312.9828 g/mol