Learn More
Alfa Aesar™ 2-Chloro-N-(2-fluoro-5-methylphenyl)benzamide, 97%
Brand:  Alfa Aesar™ H59430.03
Additional Details : CAS Number : 923742-88-5 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15485748
  • 6228.40 SEK / 1g
Estimated Shipment date
from Supplier 08-05-2018
Add to basket
Specifications
Chemical Identifiers
2-Chloro-N-(2-fluoro-5-methylphenyl)benzamide
923742-88-5
263.7
MFCD07813526
Specifications
2-Chloro-N-(2-fluoro-5-methylphenyl)benzamide
1g
Solid
263.7
923742-88-5
Off-white
97%
MFCD07813526